Skip to Content
Merck
All Photos(1)

Key Documents

21218

Sigma-Aldrich

Calcium phosphate

purum p.a., ≥96.0% (calc. as Ca3(PO4)2, KT)

Synonym(s):

β-Calcium phosphate tribasic, β-Tricalcium phosphate, tert-Calcium phosphate, tri-Calcium (ortho)phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ca3O8P2
CAS Number:
Molecular Weight:
310.18
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Assay

≥96.0% (calc. as Ca3(PO4)2, KT)

form

powder

loss

≤0.1% loss on ignition, 800 °C

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤200 mg/kg
K: ≤500 mg/kg
Na: ≤1000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/3Ca.2H3O4P/c;;;2*1-5(2,3)4/h;;;2*(H3,1,2,3,4)/q3*+2;;/p-6

InChI key

QORWJWZARLRLPR-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Calcium phosphate (tri calcium phosphate) is a colorless amorphous solid. Its pure form can be obtained by melting CaO with phosphorus (V) oxide at 1100°C. It is used as polish in toothpastes and also in making enamels and opaque glasses.
Calcium phosphate occurs naturally as the minerals: oxydapatit, voelicherite and whitlockite.[1] It is mainly used for developing osteoconductive biomaterials for use as bone substitutes.[2][3]

Application

Composite of calcium phosphate and a bioactive glass may be employed for the manufacture of customized implants, via 3D-printing process.[4] Dense and homogeneous calcium phosphate coatings may be used for coating the metal implants.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium phosphate-based osteoinductive materials.
LeGeros RZ.
Chemical Reviews, 108(11), 4742-4753 (2008)
Calcium phosphates compounds in conjunction with hydrogel as carrier for BMP-2: a study on ectopic bone formation in rats.
Hulsart-Billstrom G, et al.
Acta Biomaterialia, 7(8), 3042-3049 (2011)
S Leeuwenburgh et al.
Journal of biomedical materials research, 56(2), 208-215 (2001-05-08)
A new biomimetic method for coating metal implants enables the fast formation of dense and homogeneous calcium phosphate coatings. Titanium alloy (Ti6Al4V) disks were coated with a thin, carbonated, amorphous calcium phosphate (ACP) by immersion in a saturated solution of
3D printing of bone substitute implants using calcium phosphate and bioactive glasses.
Bergmann C, et al.
J. Eur. Ceram. Soc., 30(12), 2563-2563 (2010)
Françoise Borcard et al.
Journal of medicinal chemistry, 55(18), 7988-7997 (2012-08-18)
To control the selective adhesion of human endothelial cells and human serum proteins to bioceramics of different compositions, a multifunctional ligand containing a cyclic arginine-glycine-aspartate (RGD) peptide, a tetraethylene glycol spacer, and a gallate moiety was designed, synthesized, and characterized.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service