773956
Barium phosphate
powder, -200 mesh, 99.9% trace metals basis
Synonym(s):
Barium orthophosphate, Tribarium diphosphate
Select a Size
About This Item
Recommended Products
Quality Level
Assay
99.9% trace metals basis
form
powder
particle size
-200 mesh
mp
>300 °C
SMILES string
[Ba++].[Ba++].[Ba++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
InChI
1S/3Ba.2H3O4P/c;;;2*1-5(2,3)4/h;;;2*(H3,1,2,3,4)/q3*+2;;/p-6
InChI key
WAKZZMMCDILMEF-UHFFFAOYSA-H
Related Categories
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service