Skip to Content
Merck
All Photos(4)

Key Documents

202193

Sigma-Aldrich

Cobalt(II) carbonate hydrate

Co 43-47 %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCO3·xH2O
CAS Number:
Molecular Weight:
118.94 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
synthesis grade
form:
powder

grade

synthesis grade

Quality Level

form

powder

composition

Co, 43-47%

concentration

43.0-47.0% Co (EDTA titration)

density

4.13 g/mL at 25 °C (lit.)

SMILES string

O.[Co++].[O-]C([O-])=O

InChI

1S/CH2O3.Co.H2O/c2-1(3)4;;/h(H2,2,3,4);;1H2/q;+2;/p-2

InChI key

JLAIPADPFFTYLP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Cobalt(II) carbonate hydrate can be used as a reagent to synthesize transition metal dipicolinates by reacting with dipicolinic acid and 4,4′-bipyridine. It can also be used as a precursor to prepare Co3O4/MgO and Co3O4/Al2O3 catalysts by using MgO or Al2O3, which can be used for the hydrogenation of cinnamaldehyde to cinnamyl alcohol.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-activity MgO-supported CoMo hydrodesulfurization catalysts prepared by non-aqueous impregnation.
Kaluza L, et al.
Applied Catalysis. B, Environmental, 162, 430-436 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service