Skip to Content
Merck
All Photos(1)

Key Documents

185302

Sigma-Aldrich

Diisopropyl ether

ReagentPlus®, 99%, contains either BHT or hydroquinone as stabilizer

Synonym(s):

Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.07
bp:
68-69 °C (lit.)
vapor pressure:
120 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.5 (vs air)

Quality Level

vapor pressure

120 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

827 °F

expl. lim.

1-21 %, 100 °F

dilution

(for general lab use)

refractive index

n20/D 1.367 (lit.)

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

density

0.725 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(C)C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisopropyl ether is a colorless liquid. It is widely used as a solvent for the isolation of fats and oils. It can be synthesized from isopropyl alcohol. Its mixture with isooctane has been added to the gasoline fuel as an antiknocking agent.[1] Its synthesis from propylene and water in a single-pass trickle-bed reactor in the presence of an ion-exchange resin catalyst Amberlyst 15 has been proposed.[2] Its efficacy as a fuel additive has been reported. When released in the atmosphere, it undergoes photodecomposition by reacting with OH radicals. Kinetics and mechanism of its reaction in atmosphere have been reported.[3]

Application

Diisopropyl ether may be employed as a solvent for the preparation of aliphatic and aromatic cyanohydrins.[4]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atmospheric chemistry of automotive fuel additives: diisopropyl ether.
Wallington TJ, et al.
Environmental Science & Technology, 27(1), 98-104 (1993)
Single stage synthesis of diisopropyl ether-an alternative octane enhancer for lead-free petrol.
Heese FP, et al.
Catalysis Today, 49(1), 327-335 (1999)
Eagleson M.
Concise Encyclopedia Chemistry, 325-325 (1994)
Novel (R)-oxynitrilase sources for the synthesis of (R)-cyanohydrins in diisopropyl ether.
Kiljunen E and Kanerva LT.
Tetrahedron Asymmetry, 8(8), 1225-1234 (1997)
P Adlercreutz
Biochimica et biophysica acta, 1163(2), 144-148 (1993-05-13)
Horse liver alcohol dehydrogenase and alpha-chymotrypsin were deposited on a porous support material, Celite. After equilibration at a well-defined water activity, the catalytic activity was measured with diisopropyl ether as reaction medium. The effects of the presence of polyols and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service