Skip to Content
Merck

1.03893

SAFC

Benzenesulfonic acid

EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

Benzene sulphonic acid, Benzenesulphonic acid, Besylic acid, Phenylsulfonic acid, Phenylsulphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO3H
CAS Number:
Molecular Weight:
158.18
Beilstein:
742513
EC Number:
MDL number:
UNSPSC Code:
12352005

biological source

synthetic

Quality Level

product line

EMPROVE® EXPERT

form

crystalline

technique(s)

API processing | cocrystal formation: suitable
API processing | salt formation: suitable

impurities

endotoxins, low

application(s)

liquid formulation
parenterals
pharmaceutical

SMILES string

OS(=O)(=O)c1ccccc1

InChI

1S/C6H6O3S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H,(H,7,8,9)

InChI key

SRSXLGNVWSONIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For protein-based therapeutics, such as monoclonal antibodies (mAbs) or plasma derived proteins, often highly concentrated protein formulations are required to meet intended dosing regimens. However, this usually comes with elevated viscosities and presents a challenge in the manufacturing process and final formulation. Our Viscosity Reduction Excipient Platform reduces the viscosity and helps overcome challenges associated with highly concentrated protein solutions.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation.
Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

The Viscosity Reduction Platform is based on the combination of two new excipients, one amino acid component with one anionic component, that together can better reduce protein viscosity beyond levels achieved with single excipients. Also, combining the two components enables a balance of viscosity reduction and protein stabilization. Benzenesulfonic Acid Emprove® Expert is part of this new proprietary technology platform.

Packaging

1038930500 / 500g / Glass bottle
1038931000 / 1kg / Glass bottle

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service