Skip to Content
Merck
All Photos(1)

Key Documents

Y0000561

Vecuronium bromide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-[(2β,3α,5α,16β,17β)-3,17-Bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methyl-piperidinium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H57BrN2O4
CAS Number:
Molecular Weight:
637.73
Beilstein:
4837308
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

vecuronium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3C[C@@H]([C@@H]4OC(C)=O)[N+]5(C)CCCCC5)[C@@]2(C)C[C@@H]1N6CCCCC6

InChI

1S/C34H57N2O4.BrH/c1-23(37)39-31-20-25-12-13-26-27(34(25,4)22-29(31)35-16-8-6-9-17-35)14-15-33(3)28(26)21-30(32(33)40-24(2)38)36(5)18-10-7-11-19-36;/h25-32H,6-22H2,1-5H3;1H/q+1;/p-1/t25-,26+,27-,28-,29-,30-,31-,32-,33-,34-;/m0./s1

InChI key

VEPSYABRBFXYIB-PWXDFCLTSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vecuronium bromide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Vega-Villa et al.
British journal of anaesthesia, 112(2), 304-310 (2013-09-27)
Burn patients develop resistance to non-depolarizing neuromuscular blocking agents (NDNMBAs) and require a significantly large dose to produce a desired clinical response. Pathophysiological changes related to burn injury may alter pharmacokinetics (PK) and pharmacodynamics of NDNMBAs. The purpose of this
K Nitahara et al.
Anaesthesia, 68(9), 904-907 (2013-06-25)
We investigated whether a bolus injection of 20 ml saline with arm elevation might shorten the onset time of vecuronium administered via a dorsal hand vein. Thirty patients were randomly allocated to the bolus saline group or control group. General
L Liu et al.
British journal of anaesthesia, 112(1), 159-168 (2013-08-02)
Resistance to non-depolarizing neuromuscular blocking agents induced by sepsis is associated with the qualitative change in the nicotinic acetylcholine receptor (nAChR). This study aims to investigate the effects of sepsis on the neuromuscular block properties of vecuronium in relation to
Manuel Martin-Flores et al.
Veterinary anaesthesia and analgesia, 39(4), 385-389 (2012-05-31)
To evaluate the potency and duration of three subparalyzing doses of vecuronium (VEC) in isoflurane-anesthetized horses. Prospective experimental study. Thirteen healthy adult horses undergoing arthroscopic surgery. During isoflurane anesthesia, horses received one of three doses of vecuronium (25, 50, or
Louise Clark et al.
Veterinary anaesthesia and analgesia, 39(5), 472-479 (2012-05-31)
To compare the duration of action of vecuronium in diabetic dogs with a control group. Prospective clinical study. Forty client-owned diabetic (n = 20) and non-diabetic dogs. Dogs were considered free from other concurrent disease based on clinical examination and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service