Skip to Content
Merck
All Photos(1)

Key Documents

Y0000453

Oxycodone impurity D

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

14-Hydroxycodeinone, (5α)-7,8-Didehydro-4,5-epoxy-14-hydroxy-3-methoxy-17-methylmorphinan-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NO4
CAS Number:
Molecular Weight:
313.35
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

oxycodone

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N1([C@H]2[C@]3([C@@]4(C(Oc5c4c(ccc5OC)C2)C(=O)C=C3)CC1)O)C

InChI

1S/C18H19NO4/c1-19-8-7-17-14-10-3-4-12(22-2)15(14)23-16(17)11(20)5-6-18(17,21)13(19)9-10/h3-6,13,16,21H,7-9H2,1-2H3/t13-,16?,17+,18-/m1/s1

InChI key

YYCRAERBSFHMPL-FSFXSCMFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxycodone impurity D EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Szentesi et al.
Analytical and bioanalytical chemistry, 374(3), 427-431 (2002-10-10)
A negative Cotton effect is observed in the circular dichroism (CD) spectra of 6-oxo-morphinans in the wavelength range of n-pi* electron transitions. 6-oxo-Morphinans can be transformed into oxime derivatives with hydroxylamine and after oxime formation the CD spectra are significantly
D L Lister et al.
FEMS microbiology letters, 181(1), 137-144 (1999-11-24)
A biotransformation mixture which contained codeine and washed cells of Pseudomonas putida M10 gave rise to a number of transformation products that are of clinical importance which included hydrocodone, dihydrocodeine and 14beta-hydroxycodeine. Incubations with the same organism and codeinone gave
D López et al.
The Journal of organic chemistry, 65(15), 4671-4678 (2000-08-26)
The photooxidation of thebaine (3) with a sun lamp affords two main products: hydrodibenzofuran 10 (major) and benzofuran 11 (minor). The latter compound becomes predominant if a Hg lamp is used instead of a sun lamp. The formation of 10

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service