Skip to Content
Merck
All Photos(1)

Documents

T1985000

Trichlorotrifluoroethane

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,1,2-Trichloro-1,2,2-trifluoroethane, 1,1,2-Trichloro-trifluoroethane, Genetron® 113

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCF2CCl2F
CAS Number:
Molecular Weight:
187.38
Beilstein:
1740335
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

5.5 psi ( 20 °C)

API family

halothane

autoignition temp.

1256 °F

manufacturer/tradename

EDQM

refractive index

n20/D 1.358 (lit.)

bp

47-48 °C (lit.)

mp

−35 °C (lit.)

density

1.57 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(Cl)C(F)(Cl)Cl

InChI

1S/C2Cl3F3/c3-1(4,6)2(5,7)8

InChI key

AJDIZQLSFPQPEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trichlorotrifluoroethane EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Legal Information

Genetron is a registered trademark of Honeywell International Inc.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Ozone 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Arnoldi et al.
PloS one, 9(4), e95197-e95197 (2014-04-17)
Replication of many RNA viruses benefits from subversion of the autophagic pathway through many different mechanisms. Rotavirus, the main etiologic agent of pediatric gastroenteritis worldwide, has been recently described to induce accumulation of autophagosomes as a mean for targeting viral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service