Skip to Content
Merck
All Photos(1)

Key Documents

CRM46904

Supelco

cis-11-Vaccenic acid methyl ester

certified reference material, 10 mg/mL in heptane, ampule of 1 mL

Synonym(s):

cis-11-Octadecenoic methyl ester, cis-Vaccenic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€50.00

€50.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€50.00

About This Item

Linear Formula:
CH3(CH2)5CH=CH(CH2)9CO2CH3
CAS Number:
Molecular Weight:
296.49
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

€50.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in heptane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

single component solution

functional group

ester

shipped in

ambient

storage temp.

-10 to -25°C

SMILES string

O(C)C(=O)CCCCCCCCC\C=C\CCCCCC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h8-9H,3-7,10-18H2,1-2H3/b9-8+

InChI key

PVVODBCDJBGMJL-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-3.99 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service