Skip to Content
Merck
All Photos(1)

Key Documents

T1200000

Thiopental

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Thiopental, 5-Ethyl-5-(1-methylbutyl)-2-thiobarbituric acid, Pentothal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N2O2S
CAS Number:
Molecular Weight:
242.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

API family

thiopental

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCC(C)C1(CC)C(=O)NC(=S)NC1=O

InChI

1S/C11H18N2O2S/c1-4-6-7(3)11(5-2)8(14)12-10(16)13-9(11)15/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16)

InChI key

IUJDSEJGGMCXSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thiopental EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tark Purtuloğlu et al.
The journal of ECT, 29(1), 37-40 (2013-02-05)
To compare propofol and sodium thiopental as anesthetic agents for electroconvulsive therapy (ECT) in major depression with respect to clinical effect. Participants were composed of 96 patients with depression who were administered either propofol or sodium thiopental as an anesthetic
Bożena Bądzyńska et al.
Pharmacological reports : PR, 66(5), 856-861 (2014-08-26)
Activation of angiotensin AT2 receptors (AT2R) counteracts vasoconstrictor effects of AT1R stimulation and contributes to blood pressure control. We examined effects on mean arterial pressure (MAP) and renal hemodynamics of LKP, a tripeptide fragment of novokinine, an established AT2R agonist.
Chadi G Abdallah et al.
The journal of ECT, 28(3), 157-161 (2012-08-01)
Studies now provide strong evidence that the N-methyl-D-aspartate receptor antagonist ketamine possesses rapidly acting antidepressant properties. This study aimed to determine if a low dose of ketamine could be used to expedite and augment the antidepressant effects of electroconvulsive therapy
Cecilia Lindgren et al.
Neurocritical care, 17(3), 367-373 (2012-08-31)
Non-convulsive seizures (NCSZ) can be more prevalent than previously recognized among comatose neuro-intensive care patients. The aim of this study was to evaluate the frequency of NCSZ and non-convulsive status epilepticus (NCSE) in sedated and ventilated subarachnoid hemorrhage (SAH) patients.
H Murdoch et al.
International journal of obstetric anesthesia, 22(1), 31-35 (2012-11-06)
A national survey of current practice and preferred drug choices for both induction and maintenance of general anaesthesia for caesarean section was undertaken. Following approval by the Obstetric Anaesthetists' Association, all UK consultant members were invited to respond to an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service