Skip to Content
Merck
All Photos(1)

Key Documents

STS0015

Sigma-Aldrich

ECOSURF EH-3

greener alternative

Non-ionic surfactant

Synonym(s):

2-Ethyl hexanol EO-PO nonionic surfactant

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€37.30
500 ML
€49.40

€37.30


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€37.30
500 ML
€49.40

About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.28

€37.30


Estimated to ship on01 April 2025


Request a Bulk Order

grade

laboratory grade

Quality Level

description

non-ionic

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

480 ppm (25°C)

viscosity

25.143 cSt(40 °C)

transition temp

pour point -21 °C
cloud point 52 °C (10 wt% actives aq solution)

solubility

chlorinated solvents: soluble (and most polar organic solvents)
water: freely soluble

density

0.9651 g/mL at 40 °C

HLB

7.9

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

Legal Information

ECOSURF is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service