Skip to Content
Merck
All Photos(1)

Documents

S7135

Sigma-Aldrich

Span® 85

Synonym(s):

Sorbitane trioleate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

mol wt

957.52 g/mol

solubility

chloroform: 50 mg/mL, clear, faintly to light yellow

HLB

1.8

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@@H](O)C1OC[C@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C60H108O8/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-56(62)65-52-54(61)59-60(68-58(64)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)55(53-66-59)67-57(63)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54-55,59-61H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27+/t54-,55+,59-,60-/m1/s1

InChI key

ZBNRGEMZNWHCGA-WECPCUOASA-N

General description

Sorbitane trioleate, or Span 85, is a non-ionic detergent.

Application

Sorbitane trioleate, or Span 85, has been used in a study to develop a novel bottom-up process to produce nanoparticles containing protein and peptide for suspension in hydrofluoroalkane propellants. It has also been used in a study to analyze the application of liquid chromatography in polymer non-ionic antistatic additives.

Other Notes

Fatty acid composition: Oleic acid (C18:1) ≥ 60%; linoleic acid (C18:2) 3 - 12%; linolenic acid (C18:2) ≤ 3%; palmitoleic acid (C16:1) 2 - 6%; balance primarily palmitic acid (C16:0).

Legal Information

Span is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

437.0 °F

Flash Point(C)

225 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel bottom-up process to produce nanoparticles containing protein and peptide for suspension in hydrofluoroalkane propellants
Tan, Y., et al.
International Journal of Pharmaceutics, 413, 7-7 (2011)
M Victoria González-Rodríguez et al.
Journal of separation science, 33(22), 3595-3603 (2010-10-12)
This article investigates the applicability of HPLC-UV, ultra performance LC-evaporative light-scattering detection (UPLC-ELSD), HPLC-ESI(+)-MS and HPLC-hybrid linear ion trap (LTQ) Orbitrap MS for the analysis of different non-ionic antistatic additives, Span 20, Span 60, Span 65, Span 80, Span 85
A N Hassan et al.
International journal of food microbiology, 87(1-2), 145-152 (2003-08-21)
The influence of surfactant hydrophobicity on detachment of Escherichia coli O157:H7 from lettuce was determined. Lettuce pieces inoculated with the pathogen were rinsed with Tween and Span surfactants of different hydrophobicity. Of the Tweens, only Tween 85, the Tween with
Hiroaki Todo et al.
International journal of pharmaceutics, 271(1-2), 41-52 (2004-05-08)
The effect of pulmonary absorption enhancers on the stability of active ingredients is an important factor for successful inhalation therapy as well as the effect on pharmacological activity and safety. We examined the effect of pulmonary absorption enhancers on the
P M Blackett et al.
Pharmaceutical research, 12(11), 1689-1693 (1995-11-01)
The purpose of the work is to study the adsorption of Oleic acid and Span 85 (materials frequently used in aerosols as surfactants) onto partially amorphous and essentially crystalline salbutamol sulphate, attempting to understand the behaviour of metered dose inhalers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service