Skip to Content
Merck
All Photos(1)

Documents

STS0013

Sigma-Aldrich

TERGITOL 15-S-5

greener alternative

Non-ionic surfactant

Synonym(s):

Alcohols C6-C17 (secondary) poly (3-6) ethoxylates, Secondary alcohol ethoxylate 5 EO

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.28

grade

laboratory grade

Quality Level

description

non-ionic

form

liquid

mol wt

average mol wt 420 g/mol by calculation

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

viscosity

35 cP(25 °C)

transition temp

pour point -25 °C

solubility

water: dispersible (visual)

density

0.963 g/mL at 20 °C

HLB

10.5

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".
Tergitol 15-S-5 is a secondary alcohol ethoxylate with 5 ethylene oxide units

Application


  • Spreading on and penetration into thin, permeable print media: application to ink-jet printing.: This study explores the dynamics of spreading and penetration of liquids on thin, permeable media, focusing on applications in ink-jet printing. The research addresses the role of surfactants, like TERGITOL 15-S-5, in modifying the interfacial properties to optimize the print quality and efficiency (Daniel RC, et al., 2006).

Features and Benefits

  • Excellent emulsifier & detergent
  • Aids in rinseability of solvent-based systems

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service