Skip to Content
Merck
All Photos(1)

Key Documents

S6689

Sigma-Aldrich

SynaptoRed C2

≥95% purity (HPLC), solid

Synonym(s):

FM4-64, N-(3-Triethylammoniopropyl)-4-(6-(4-(diethylamino)phenyl) hexatrienyl)pyridinium dibromide, Pyridinium, 4-[6-[4-(diethylamino)phenyl]- 1,3,5-hexatrienyl]-1-[3-(triethylammonio) propyl]-, dibromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€636.00

€636.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€636.00

About This Item

Empirical Formula (Hill Notation):
C30H45Br2N3
CAS Number:
Molecular Weight:
607.51
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€636.00


Estimated to ship on31 May 2025


Request a Bulk Order

Product Name

SynaptoRed C2, ≥95% (HPLC), solid

Quality Level

Assay

≥95% (HPLC)

form

solid

technique(s)

titration: suitable

color

deep purple

solubility

DMSO: 10 mg/mL

εmax

≥37 at 538-548 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[Br-].[Br-].CCN(CC)c1ccc(/C=C\C=C\C=C/c2cc[n+](CCC[N+](CC)(CC)CC)cc2)cc1

InChI

1S/C30H45N3.2BrH/c1-6-32(7-2)30-20-18-28(19-21-30)16-13-11-12-14-17-29-22-25-31(26-23-29)24-15-27-33(8-3,9-4)10-5;;/h11-14,16-23,25-26H,6-10,15,24,27H2,1-5H3;2*1H/q+2;;/p-2

InChI key

AFVSZGYRRUMOFH-UHFFFAOYSA-L

General description

SynaptoRed C2 is an amphiphilic styryl dye.[1] It stains endocytic intermediates and vacuole membranes.[2]

Application

SynaptoRed C2 has been used for propidium iodide (PI) blockage.[3]

Biochem/physiol Actions

Fluorescent dye suitable for monitoring synaptic activity at neuromuscular junctions.

Legal Information

SynaptoRed is a trademark of Biotium, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena Plotka et al.
Viruses, 11(7) (2019-07-22)
Bacteria that thrive in extreme conditions and the bacteriophages that infect them are sources of valuable enzymes resistant to denaturation at high temperatures. Many of these heat-stable proteins are useful for biotechnological applications; nevertheless, none have been utilized as antibacterial
Plant Cell Expansion: Methods and Protocols (2014)
Construction of a functional Casparian strip in non-endodermal lineages is orchestrated by two parallel signaling systems in Arabidopsis thaliana
Li P, et al.
Current Biology, 28(17), 2777-2786 (2018)
B Wendland et al.
The Journal of cell biology, 135(6 Pt 1), 1485-1500 (1996-12-01)
A complete understanding of the molecular mechanisms of endocytosis requires the discovery and characterization of the protein machinery that mediates this aspect of membrane trafficking. A novel genetic screen was used to identify yeast mutants defective in internalization of bulk
Biology of the Fungal Cell (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service