Skip to Content
Merck
All Photos(1)

Key Documents

PHL83276

Aurantio-obtusin

phyproof® Reference Substance

Synonym(s):

1,3,7-Trihydroxy-2,8-dimethoxy-6-methyl-9,10-anthracenedione

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€413.00

€413.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€413.00

About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€413.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O(C)c1c2c(cc(c1O)C)C(=O)c3c(c(c(c(c3)O)OC)O)C2=O

InChI

1S/C17H14O7/c1-6-4-7-11(17(24-3)12(6)19)14(21)10-8(13(7)20)5-9(18)16(23-2)15(10)22/h4-5,18-19,22H,1-3H3

InChI key

RNXZPKOEJUFJON-UHFFFAOYSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang-Ho Kang et al.
PloS one, 15(5), e0225564-e0225564 (2020-05-08)
Senna tora is an annual herb with rich source of anthraquinones that have tremendous pharmacological properties. However, there is little mention of genetic information for this species, especially regarding the biosynthetic pathways of anthraquinones. To understand the key genes and
Eun Kyeong Lee et al.
Molecules (Basel, Switzerland), 25(10) (2020-05-28)
The purpose of this study was to determine the antiallergic effects of AF-343, a mixture of natural plant extracts from Cassia tora L., Ulmus pumila L., and Taraxacum officinale, on rat basophilic leukemia (RBL-2H3) cells. The inhibitory effects on cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service