Recommended Products
CofA
current certificate can be downloaded
packaging
ampule of 1.2 mL
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
matrix material
storage temp.
2-8°C
General description
Application
Analysis Note
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
WGK
""
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service