Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
50 mg | Check Cart for Availability | € 554,00 |
About This Item
Quality Segment
grade
certified reference material, pharmaceutical secondary standard
agency
traceable to USP 1096735
API family
carmustine
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
application(s)
pharmaceutical
storage temp.
-10 to -25°C
SMILES string
ClCCNC(=O)NCCCl
InChI
1S/C5H10Cl2N2O/c6-1-3-8-5(10)9-4-2-7/h1-4H2,(H2,8,9,10)
InChI key
VBWBRZHAGLZNST-UHFFFAOYSA-N
General description
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description pharmaceutical secondary standard, certified reference material | description certified reference material, pharmaceutical secondary standard | description pharmaceutical secondary standard, certified reference material | description pharmaceutical secondary standard, certified reference material |
| grade certified reference material, pharmaceutical secondary standard | grade certified reference material, pharmaceutical secondary standard | grade certified reference material, pharmaceutical secondary standard | grade certified reference material, pharmaceutical secondary standard |
| application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| packaging pkg of 50 mg | packaging pkg of 50 mg | packaging pkg of 50 mg | packaging pkg of 40 mg |
| storage temp. -10 to -25°C | storage temp. 2-30°C | storage temp. 2-8°C | storage temp. 2-30°C |
| API family carmustine | API family bendamustine | API family lomustine | API family triclocarban |
Still not finding the right product?
Explore all of our products under Carmustine Related Compound A
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



