Skip to Content
Merck
All Photos(1)

Key Documents

PHR3149

Supelco

Triclocarban Related Compound C

pharmaceutical secondary standard, certified reference material

Synonym(s):

1,3-Bis(3,4-dichlorophenyl)urea, 3,3′,4,4′-Tetrachlorocarbanilide

Sign Into View Organizational & Contract Pricing

Select a Size

40 MG
€452.00

€452.00


Estimated to ship on04 May 2025


Request a Bulk Order

Select a Size

Change View
40 MG
€452.00

About This Item

Empirical Formula (Hill Notation):
C13H8Cl4N2O
CAS Number:
Molecular Weight:
350.03
UNSPSC Code:
41116107
NACRES:
NA.24

€452.00


Estimated to ship on04 May 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1681845

API family

triclocarban

CofA

current certificate can be downloaded

packaging

pkg of 40 mg

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)NC(=O)Nc2cc(c(cc2)Cl)Cl)Cl

InChI

1S/C13H8Cl4N2O/c14-9-3-1-7(5-11(9)16)18-13(20)19-8-2-4-10(15)12(17)6-8/h1-6H,(H2,18,19,20)

InChI key

ZDPIZPXVHVYTOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6959(changes by product) in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service