Skip to Content
Merck
All Photos(1)

Key Documents

PHR2806

Supelco

Mupirocin

pharmaceutical secondary standard

Synonym(s):

Mupirocin, 5,9-Anhydro-2,3,4,8-tetradeoxy-8-[[3-(2-hydroxy-1-methylpropyl)oxiranyl]methyl]-3-methyl-[2E,8[2S,3S(1S,2S)]]-L-talonon-2-enonic acid 8-carboxyoctyl ester, BRL 4910A, Pseudomonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€157.00

€157.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€157.00

About This Item

Empirical Formula (Hill Notation):
C26H44O9
CAS Number:
Molecular Weight:
500.62
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€157.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1448901

API family

mupirocin

packaging

pkg of 100 mg

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

[H][C@@]1(CO[C@@H](C\C(C)=C\C(=O)OCCCCCCCCC(O)=O)[C@H](O)[C@@H]1O)C[C@@H]2O[C@@]2([H])[C@@H](C)[C@H](C)O

InChI

1S/C26H44O9/c1-16(13-23(30)33-11-9-7-5-4-6-8-10-22(28)29)12-20-25(32)24(31)19(15-34-20)14-21-26(35-21)17(2)18(3)27/h13,17-21,24-27,31-32H,4-12,14-15H2,1-3H3,(H,28,29)/b16-13+/t17-,18-,19-,20-,21-,24+,25-,26-/m0/s1

InChI key

MINDHVHHQZYEEK-HBBNESRFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Antibiotic; inhibits isoleucyl-tRNA synthetase (IRS).

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC8244 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. What is the purity of PHR2806? I'm considering its use as a selective agent in microbiological media.

    1 answer
    1. The certification for this product does not include a purity specification. The certified purity for individual lots is available on the respective certificates of analysis in the documents section of the product page. This product is intended for testing according to relevant compendial monographs and has not been evaluated for efficacy in other applications. This product is not sterile.

      Helpful?

  2. What are the storage conditions for Mupirocin PHR2806 (CAS 12650-69-0)?

    1 answer
    1. As listed in the 'PROPERTIES' section, the recommended storage temperature for this product is -10 to -25 °C.

      Helpful?

  3. What is the solubility of the product Mupirocin PHR2806 (CAS 12650-69-0)?

    1 answer
    1. The product is soluble in DMSO, with solubility ranging approximately from 20 mg/mL to 100 mg/mL, and in ethanol at approximately 5 mg/mL. However, it is insoluble in water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service