Skip to Content
Merck
All Photos(1)

Documents

PHR1599

Supelco

Alendronate Sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Alendronate sodium trihydrate, 4-amino-1-hydroxy-1-phosphonobutyl phosphonic acid, monosodium, MK-217

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12NaNO7P2 · 3 H2O
CAS Number:
Molecular Weight:
325.12
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 830
traceable to Ph. Eur. Y0001727
traceable to USP 1012780

API family

alendronate

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Na+].[H]O[H].[H]O[H].[H]O[H].NCCCC(O)(P(O)(O)=O)P(O)([O-])=O

InChI

1S/C4H13NO7P2.Na.3H2O/c5-3-1-2-4(6,13(7,8)9)14(10,11)12;;;;/h6H,1-3,5H2,(H2,7,8,9)(H2,10,11,12);;3*1H2/q;+1;;;/p-1

InChI key

DCSBSVSZJRSITC-UHFFFAOYSA-M

Gene Information

human ... FDPS(2224)

Looking for similar products? Visit Product Comparison Guide

General description

Alendronate Sodium, an aminobisphosphonate compound, has been effectively used for the treatment of osteolysis, Paget′s disease and osteoporosis.
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

Alendronate Sodium may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and plasma samples by high-performance liquid chromatography method.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Alendronate sodium trihydrate is a bone resorption inhibitor; farnesyl diphosphate synthase inhibitor (IC50 = 460 nM); CD45 protein tyrosine phosphatase inhibitor.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2083 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of alendronate sodium by ion chromatography with refractive index detection.
Han YHR and Qin XZ
Journal of Chromatography A, 719(2), 345-352 (1996)
High-performance liquid chromatography method for determining alendronate sodium in human plasma by detecting fluorescence: application to a pharmacokinetic study in humans.
Yun MH, et al.
Journal of Pharmaceutical and Biomedical Analysis, 40(1), 168-172 (2006)
A fast and simple spectrofluorometric method for the determination of alendronate sodium in pharmaceuticals.
Ezzati JND, et al.
BioImpacts, 4(1), 39-42 (2014)
Hüseyin Abdik et al.
Molecular biology reports, 46(1), 763-776 (2018-12-07)
Bisphosphonate-induced osteonecrosis of the jaw (BIONJ) is a commonly encountered side effect of Bisphosphonates (BPs). Although certain aspects of BIONJ have been studied, the effects of BPs on the proliferation, differentiation, and maintenance of dental stem cells (DSC) in way

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service