Skip to Content
Merck
All Photos(1)

Key Documents

45521

Supelco

Glyphosate

PESTANAL®, analytical standard

Synonym(s):

N-(Phosphonomethyl)glycine, Glyphosate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2P(O)CH2NHCH2CO2H
CAS Number:
Molecular Weight:
169.07
Beilstein:
2045054
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

230 °C (dec.) (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)CNCP(O)(O)=O

InChI

1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)

InChI key

XDDAORKBJWWYJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glyphosate is a nonselective, broad-spectrum, perennial herbicide, employed at post-emergence stage for weed and vegetation control.

Application

Glyphosate may be used as an analytical reference standard for the quantification of the analyte in environmental samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of glyphosate by ion chromatography
Zhu Y, et al.
Talanta, 850(1-2), 297-301 (1999)
Glyphosate: A Unique Global Herbicide (1997)
Re-evaluation of glyphosate determination in water by liquid chromatography coupled to electrospray tandem mass spectrometry
Ibanez M, et al.
Journal of Chromatography A, 1134(1-2), 51-55 (2006)
Remy Babich et al.
Environment international, 144, 106019-106019 (2020-08-21)
Chronic kidney disease of unknown etiology (CKDu) is an emerging global concern affecting several agricultural communities in the Americas and South Asia. Environmental contaminants such as heavy metals (e.g., Cd, As, Pb, and V) and organic pesticides (e.g., glyphosate) in
Stephen O Duke
Journal of agricultural and food chemistry, 59(11), 5835-5841 (2010-10-06)
High levels of aminomethylphosphonic acid (AMPA), the main glyphosate metabolite, have been found in glyphosate-treated, glyphosate-resistant (GR) soybean, apparently due to plant glyphosate oxidoreductase (GOX)-like activity. AMPA is mildly phytotoxic, and under some conditions the AMPA accumulating in GR soybean

Articles

Glyphosate and related compounds are measured in oatmeal and infant cereal using ion-exchange polymer-based particles for HPLC and SPE. Low level detection was obtained.

Glyphosate analysis: LC-MS/MS method with Supel™ Carbon LC U/HPLC column for stability and retention.

Protocols

EPA Method 547 outlines the analysis of glyphosate in drinking water by direct aqueous injection HPLC, post column derivatization, and fluorescence detection

LC/MS Analysis of Glyphosate and Metabolites on apHera™ NH2, 2 mm I.D. Column

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service