Skip to Content
Merck
All Photos(1)

Key Documents

PHR1303

Supelco

3,4,4` Trichlorocarbanilide (Triclocarban)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

3,4,4′-Trichlorocarbanilide, 1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea, TCC, Triclocarban

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NHCONHC6H4Cl
CAS Number:
Molecular Weight:
315.58
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

vapor pressure

<0.1 mmHg ( 25 °C)

API family

triclocarban

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

254-256 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Clc1ccc(NC(=O)Nc2ccc(Cl)c(Cl)c2)cc1

InChI

1S/C13H9Cl3N2O/c14-8-1-3-9(4-2-8)17-13(19)18-10-5-6-11(15)12(16)7-10/h1-7H,(H2,17,18,19)

InChI key

ICUTUKXCWQYESQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4,4′-Trichlorocarbanilide (Triclocarban) is an ester of 3,4,4′-trichlorocarbanilide and belongs to the group of endocrine disruptors. It is widely used as a preservative and bactericide in a variety of personal care products.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0063 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>302.0 °F

Flash Point(C)

> 150 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoliu Zhou et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 881-882, 27-33 (2011-12-24)
3,4,4'-Trichlorocarbanilide (triclocarban, TCC) is widely used as an antimicrobial agent in a variety of consumer and personal care products. Because of its widespread use, the potential for human exposure to TCC is high. Human exposure to TCC may be assessed
Simultaneous determination of parabens, triclosan and triclocarban in water by liquid chromatography/electrospray ionisation tandem mass spectrometry
Gonzalez-Marino I, et al.
Rapid Communications in Mass Spectrometry, 23(12), 1756-1766 (2009)
Arjun K Venkatesan et al.
Journal of hazardous materials, 229-230, 29-35 (2012-06-30)
The antimicrobial agents triclosan (TCS), triclocarban (TCC) and their associated transformation products are of increasing concern as environmental pollutants due to their potential adverse effects on humans and wildlife, including bioaccumulation and endocrine-disrupting activity. Analysis by tandem mass spectrometry of
Jian-Liang Zhao et al.
Water research, 47(1), 395-405 (2012-11-07)
Triclosan (TCS) and triclocarban (TCC) are two commonly used personal care products. They may enter into aquatic environments after consumption and pose potential risks to aquatic organisms. We investigated the occurrence and fate of TCS and TCC in five large
F Gressier et al.
Journal of neural transmission (Vienna, Austria : 1996), 122(1), 35-42 (2014-07-23)
The cytochrome P450 2D6 (CYP2D6) is involved in the metabolism of most antidepressants. Comedication with a potent CYP2D6 inhibitor can convert patients with extensive metabolizer (EM) or ultra-rapid metabolizer (UM) genotypes into poor metabolizer (PM) phenotypes. Since comedication is frequent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service