Skip to Content
Merck

Skip To

1499414

USP

Paricalcitol solution

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(1α,3β,7E,22E)-19-Nor-9,10-secoergosta-5,7,22-triene-1,3,25-triol solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
3 mL
Check Cart for Availability
€ 838,00

About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 838,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

paricalcitol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

O[C@H]1C[C@@H](CC(=C\C=C2\[C@H]3[C@@]([C@H](CC3)[C@H](C)\C=C\[C@@H](C(O)(C)C)C)(CCC\2)C)C1)O

InChI

1S/C27H44O3/c1-18(8-9-19(2)26(3,4)30)24-12-13-25-21(7-6-14-27(24,25)5)11-10-20-15-22(28)17-23(29)16-20/h8-11,18-19,22-25,28-30H,6-7,12-17H2,1-5H3/b9-8+,21-11+/t18-,19+,22-,23-,24-,25+,27-/m1/s1

InChI key

BPKAHTKRCLCHEA-UBFJEZKGSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Paricalcitol solution USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Paricalcitol
  • Paricalcitol Capsules
  • Paricalcitol Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
149940310863121086141
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

paricalcitol

API family

paricalcitol

API family

calcitriol

API family

calcipotriene

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

55.4 °F

flash_point_c

13 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. ¿De qué concentración es la solución? ¿En qué disolvente está?

    1 answer
    1. This item is an analytical standard solution from the United States Pharmacopeia. According to the supplier information, this is a 513 ug/mL solution in Ethanol. See the link below to review the supplier Certificate for the current lot:
      https://static.usp.org/pdf/EN/referenceStandards/certificates/1499414-R096Q0.pdf

      Helpful?

  2. Where can we find technical data for concentration and its resuspension components for this product?

    1 answer
    1. This is a reference standard from the USP. It is a 513 ug/mL solution of Paricalcitol in ethanol. Please see the links below to the USP product page as well as the Certificate for the current lot.
      USP Product page:
      https://store.usp.org/product/1499414

      USP Certificate:
      https://static.usp.org/pdf/EN/referenceStandards/certificates/1499414-R096Q0.pdf

      Helpful?

Reviews

No rating value

Active Filters