Skip to Content
Merck
All Photos(1)

Key Documents

PHR1246

Supelco

Fenofibrate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

FEN, 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid isopropyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€118.00

€118.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€118.00

About This Item

Empirical Formula (Hill Notation):
C20H21ClO4
CAS Number:
Molecular Weight:
360.83
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€118.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. F0048000
traceable to USP 1269447

API family

fenofibrate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)OC(=O)C(C)(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2

InChI

1S/C20H21ClO4/c1-13(2)24-19(23)20(3,4)25-17-11-7-15(8-12-17)18(22)14-5-9-16(21)10-6-14/h5-13H,1-4H3

InChI key

YMTINGFKWWXKFG-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Fenofibrate is a third-generation fibric acid derivative which is widely employed for the treatment of atherogenic dyslipidemia, hypertriglyceridemia, mixed dyslipidemia and hypercholesterolemia.[1]

Application

Fenofibrate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography[1][2][3] and spectrophotometry[3] techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0257 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Oral

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical studies and square-wave voltammetric determination of fenofibrate in pharmaceutical formulations
Yardimci C and Ozaltin N
Analytical and Bioanalytical Chemistry, 378(2), 495-498 (2004)
Estimation of Atorvastatin Calcium and Fenofibrate in Tablets by Derivative Spectrophotometry and Liquid Chromatography
Nakarani NV, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 90(3), 700-705 (2007)
Simultaneous Estimation of Rosuvastatin Calcium and Fenofibrate in Bulk and in Tablet Dosage Form by UV-Spectrophotometry and RP-HPLC
Karunakaran A, et al.
Stamford Journal of Pharmaceutical Sciences, 4(1), 58-63 (2011)
Jianfeng Huang et al.
Biochimica et biophysica acta, 1831(10), 1555-1565 (2013-05-01)
Obesity is associated with an increased risk for malignant lymphoma development. We used Bcr/Abl transformed B cells to determine the impact of aggressive lymphoma formation on systemic lipid mobilization and turnover. In wild-type mice, tumor size significantly correlated with depletion
Mark D Sullivan et al.
JAMA psychiatry, 70(10), 1041-1047 (2013-08-16)
Depression has been identified as a risk factor for dementia among patients with type 2 diabetes mellitus but the cognitive domains and patient groups most affected have not been identified. To determine whether comorbid depression in patients with type 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service