Skip to Content
Merck
All Photos(1)

Key Documents

PHR1197

Supelco

Iminostilbene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

5H-Dibenz[b,f]azepine, Carbamazepine Related Compound B, Iminostilbene

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€476.00

€476.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€476.00

About This Item

Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
Beilstein:
1343358
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€476.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1093023

API family

carbamazepine, oxcarbazepine 

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

196-199 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

N1c2ccccc2C=Cc3ccccc13

InChI

1S/C14H11N/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)15-13/h1-10,15H

InChI key

LCGTWRLJTMHIQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards..

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9013 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koleta Hemine et al.
Carbohydrate polymers, 250, 116957-116957 (2020-10-15)
It is widely believed that the hydrophobic effect governs the binding of guest molecules to cyclodextrins (CDs). However, it is also known that high hydrophobicity of guest molecules does not always translate to the formation of stable inclusion complexes with
S M Furst et al.
Biochemical pharmacology, 45(6), 1267-1275 (1993-03-24)
Carbamazepine is an anticonvulsant which is associated with a significant incidence of hypersensitivity reactions including agranulocytosis. We have postulated that many drug hypersensitivity reactions, especially agranulocytosis and lupus, are due to reactive metabolites generated by the myeloperoxidase (MPO) (EC 1.11.1.7)
S M Furst et al.
Drug metabolism and disposition: the biological fate of chemicals, 23(5), 590-594 (1995-05-01)
Carbamazepine therapy is associated with several types of idiosyncratic drug reactions, including hematological disorders. In previous studies, we found that carbamazepine was metabolized by the myeloperoxidase/H2O2 system of activated neutrophils, and covalent binding of the drug to neutrophils was observed.
A Varenne et al.
Journal of immunological methods, 186(2), 195-204 (1995-10-26)
As part of our ongoing work to extend the range of applications of the non-isotopic carbonyl metalloimmunoassay (CMIA), previously developed in our laboratory, we describe here the first CMIA study of carbamazepine. The CMIA method uses a metal carbonyl complex
B Moosmann et al.
Biological chemistry, 382(11), 1601-1612 (2002-01-05)
Oxidative stress is a widespread phenomenon in the pathology of neurodegenerative diseases such as Alzheimer's disease, Parkinson's disease, and amyotrophic lateral sclerosis. Neuronal cell death due to oxidative stress may causally contribute to the pathogeneses of these diseases. Therefore, neuroprotective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service