Skip to Content
Merck
All Photos(1)

Key Documents

I1308

Sigma-Aldrich

10,11-Dihydro-5H-dibenz[b,f]azepine

97%

Synonym(s):

Iminodibenzyl

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€43.50

€43.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€43.50

About This Item

Empirical Formula (Hill Notation):
C14H13N
CAS Number:
Molecular Weight:
195.26
Beilstein:
152732
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€43.50


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

105-108 °C (lit.)

SMILES string

C1Cc2ccccc2Nc3ccccc13

InChI

1S/C14H13N/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)15-13/h1-8,15H,9-10H2

InChI key

ZSMRRZONCYIFNB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Kornhuber et al.
European journal of medicinal chemistry, 44(6), 2667-2672 (2009-01-16)
We developed a structure-property-activity relationship (SPAR)-model for psychopharmacological drugs acting as non-competitive 5-HT(3A) receptor antagonists by using a decision-tree learner provided by the RapidMiner machine learning tool. A single molecular descriptor, namely the molecular dipole moment per molecular weight (mu/MW)
Yulia Kaluzhny et al.
Alternatives to laboratory animals : ATLA, 43(2), 101-127 (2015-05-23)
The 7th Amendment to the EU Cosmetics Directive and the EU REACH Regulation have reinforced the need for in vitro ocular test methods. Validated in vitro ocular toxicity tests that can predict the human response to chemicals, cosmetics and other

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service