Skip to Content
Merck
All Photos(1)

Key Documents

P9327

Sigma-Aldrich

Potassium chloride solution

0.075 M, sterile-filtered, BioXtra, suitable for cell culture

Synonym(s):

Potassium chloride for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein:
1711999
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.75
form:
solution

sterility

sterile-filtered

Quality Level

product line

BioXtra

form

solution

concentration

0.075 M

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium chloride solution is a hypotonic solution used to enlarge cells for the adequate spreading of metaphase chromosomes for karyotyping.

Application

Potassium chloride solution has been used as a hypotonic solution for resuspension of isolated pellet during the control fluorescence in situ hybridization slide preparation protocol.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calista K L Ng et al.
Human molecular genetics, 24(11), 3163-3171 (2015-02-26)
mRNA decay is an essential and active process that allows cells to continuously adapt gene expression to internal and environmental cues. There are two mRNA degradation pathways: 3' to 5' and 5' to 3'. The DCPS protein is the scavenger
Linda K Månsson et al.
Faraday discussions, 181, 49-69 (2015-05-15)
We describe a route towards self-assembled colloidal molecules, where thermoresponsive microgels serve as discrete, externally tunable interaction sites. The ability of poly(N-isopropylacrylamide) (PNIPAM) and poly(N-isopropylmethacrylamide) (PNIPMAM) microgels to adsorb to the oil/water (O/W) interface and create Pickering-stabilized mini-emulsions was first
Masatoshi Takagaki et al.
Experimental neurology, 252, 12-17 (2013-11-20)
Sedatives in the neurointensive care unit can strongly influence patients' risks of developing secondary brain damage. In particular, isoflurane, a volatile anesthetic, has been recently re-introduced to the neurointensive care unit, and first clinical studies suggest beneficial effects due to
Hanne Sørup Tastesen et al.
Amino acids, 46(7), 1659-1671 (2014-03-25)
High-protein diets induce alterations in metabolism that may prevent diet-induced obesity. However, little is known as to whether different protein sources consumed at normal levels may affect diet-induced obesity and associated co-morbidities. We fed obesity-prone male C57BL/6J mice high-fat, high-sucrose
Yuanyuan Qu et al.
Nucleic acids research, 41(10), 5263-5272 (2013-04-13)
Bacterial nucleoid-associated proteins, such as H-NS-like proteins in Enterobacteriaceae, are abundant DNA-binding proteins that function in chromosomal DNA organization and gene transcription regulation. The Mycobacterium tuberculosis Lsr2 protein has been proposed to be the first identified H-NS analogue in Gram-positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service