P2097
Potassium permanganate
meets USP testing specifications
About This Item
Recommended Products
Agency
USP/NF
meets USP testing specifications
form
crystalline
reaction suitability
reagent type: oxidant
application(s)
pharmaceutical (small molecule)
SMILES string
[K+].[O-][Mn](=O)(=O)=O
InChI
1S/K.Mn.4O/q+1;;;;;-1
InChI key
VZJVWSHVAAUDKD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation
Target Organs
Brain
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service