Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium permanganate
Synonym(s):
Potassium permanganate
Empirical Formula (Hill Notation):
KMnO
4
CAS Number:
7722-64-7
Molecular Weight:
158.03
EC Number:
231-760-3
Products
Product Category
salts (6)
Brand
Sigma-Aldrich (4)
Supelco (3)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Special Grade
ACS reagent (2)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
223468
Potassium permanganate,
ACS reagent, ≥99.0%
Expand
View Pricing
1.05082
Potassium permanganate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
399124
Potassium permanganate,
ACS reagent, ≥99.0%, low in mercury
Expand
View Pricing
238511
Potassium permanganate,
≤150 μm particle size, 97%
Expand
View Pricing
1.05080
Potassium permanganate,
cryst. EMPLURA
®
Expand
View Pricing
1.05084
Potassium permanganate,
for analysis (max. 0.000005% Hg) EMSURE
®
ACS
Expand
View Pricing
P2097
Potassium permanganate,
meets USP testing specifications
Expand
View Pricing