Skip to Content
Merck
All Photos(1)

Documents

NIST695

Trace elements in multi-nutrient fertilizer

NIST® SRM® 695

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 70 g

manufacturer/tradename

NIST®

application(s)

agriculture

format

matrix material

General description

A unit of SRM 695 consists of approximately 70 g of jet-milled fertilizer.

SRM 695_cert

SRM 695_SDS

Application

NIST695 Trace elements in multi-nutrient fertilizer standard reference material (SRM) is intended primarily for use in the evaluation of techniques employed in the analysis of multi-nutrient fertilizer materials and materials of a similar matrix.

Features and Benefits

  • SRM 695 is available with certified mass fraction values of major, minor, and trace elements.  
  • The NIST certificate is provided with source, preparation, analysis details and use instructions.

Other Notes

Certified for the analytes listed below.
Calcium (Ca), Manganese (Mn), Iron (Fe), Sodium (Na), Magnesium (Mg), Potassium (K), Zinc (Zn), Arsenic (As), Mercury (Hg), Cadmium (Cd), Molybdenum (Mo), Chromium (Cr), Nickel (Ni), Cobalt (Co), Lead (Pb), Copper (Cu), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Aluminum (Al), Boron (B), Nitrogen (N), Phosphorous (P), Selenium (Se), Chlorine (Cl), Titanium (Ti)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Flame over circleHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Ox. Sol. 3 - STOT RE 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of macronutrients, micronutrients and trace elements in mineral fertilizers by inductively coupled plasma optical emission spectrometry.
Souza O D S, et al.
Spectrochimica Acta. Part B: Atomic Spectroscopy, 96, 1-7 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service