NIST2141
Urea
NIST® SRM® 2141
Synonym(s):
Carbamide, Carbonyldiamide
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
crystals
packaging
pkg of 2 g
manufacturer/tradename
NIST®
mp
132-135 °C (lit.)
density
1.335 g/mL at 25 °C (lit.)
application(s)
clinical testing
SMILES string
NC(N)=O
InChI
1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)
InChI key
XSQUKJJJFZCRTK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Other Notes
Nitrogen (N)
See certificate for values and more details at nist.gov/SRM.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service