Skip to Content
Merck
All Photos(1)

Key Documents

M7780

Supelco

3-Methylhistamine dihydrochloride

analytical standard

Synonym(s):

3-Methyl-4-(β-aminoethyl)imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€181.00

€181.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€181.00

About This Item

Empirical Formula (Hill Notation):
C6H11N3 · 2HCl
CAS Number:
Molecular Weight:
198.09
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€181.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

Cl.Cl.Cn1cncc1CCN

InChI

1S/C6H11N3.2ClH/c1-9-5-8-4-6(9)2-3-7;;/h4-5H,2-3,7H2,1H3;2*1H

InChI key

KWEIZIDNCJBKIY-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Murray et al.
Journal of chromatography, 567(2), 289-298 (1991-07-05)
An assay has been developed for N tau-methylhistamine, a major metabolite of the autocoid histamine, based on gas chromatography-electron-capture negative-ion chemical ionisation mass spectrometry. N tau-Methylhistamine was extracted from urine by cation-exchange chromatography and converted to its di-(3,5-bistrifluoromethylbenzoyl) derivative. The
E Oosting et al.
Agents and actions, 33(1-2), 215-217 (1991-05-01)
A newly developed radioimmunoassay (RIA, Y) for the determination of urinary N tau-methylhistamine concentrations was correlated with gas chromatography mass spectrometry (GCMS, X). In 34 urine samples, with histamine and N tau-methylhistamine levels within our reference values, the correlation was:
J C Fitzpatrick et al.
The Journal of surgical research, 49(4), 293-297 (1990-10-01)
Carcinine (beta-alanylhistamine) is an imidazole dipeptide that exists in mammalian hearts, increases cardiac contractility, and is metabolically linked to carnosine (beta-alanylhistidine), a non-mast cell histidine and histamine precursor during stress. We have previously shown that tissue carnosine levels are regulated
Tibor Mikó et al.
Journal of medicinal chemistry, 46(8), 1523-1530 (2003-04-04)
In an extension of very recently published studies on successful imidazole replacements in some series of histamine H(3) receptor antagonists, we report on a new class of lipophilic nonimidazole antagonist having an aliphatic tertiary amino moiety connected to a benzyl
T Iwashina et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 48(9), 1187-1191 (1998-01-07)
In order to trace the loss of N tau-methylhistamine, a principal metabolite of histamine, during extraction and purification from human plasma and urine samples, N tau-[3H]methylhistamine was prepared in two steps from N alpha t-butoxycarbonylhistamine (II). In the first step

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service