Skip to Content
Merck
All Photos(1)

Key Documents

HVS10

Small Sample Viscosity Standard ASTM D2162

UKAS ISO/IEC17025 and ISO Guide 34 certified, kinematic viscosity 1009 cSt (40 °C)

Sign Into View Organizational & Contract Pricing

Select a Size

60 ML
€54.70

€54.70


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
60 ML
€54.70

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€54.70


Estimated to ship on07 May 2025


Request a Bulk Order

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

kinematic viscosity

1009 cSt(40 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Small Sample Viscosity Standards ASTM D2162 can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service