Skip to Content
Merck
All Photos(1)

Key Documents

VISCCS14

CCS Viscosity Standard

ASTM D 5293 and SAE Specification J300

Synonym(s):

Cold-Cranking Simulator Viscosity Standard

Sign Into View Organizational & Contract Pricing

Select a Size

14 X 500 ML
€1,470.00

€1,470.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
14 X 500 ML
€1,470.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€1,470.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

shelf life

limited shelf life, expiry date on the label

packaging

kit of 14 × 500 mL

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation. Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon CCS Viscosity Standards can be found on the Paragon Scientific webpage.

CCS Calibration Kit - 14 x 500mL, includes following products: CL10, CL12, CL14, CL16, CL19, CL22, CL25 (x2) , CL28, CL32, CL38, CL48, CL60, CL68. Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) oil testing to ASTM D5293 and SAE Specification J300.

Features and Benefits

  • Full Traceability to national standards.
  • Ensures full compliance to ASTM & IP test method protocol.
  • Shelf life of 2 years.
  • Tamper-evident security packaging.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service