Skip to Content
Merck
All Photos(1)

Key Documents

H5144

Supelco

Heroin hydrochloride solution

100 μg/mL in methanol, analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO5 · HCl
CAS Number:
Molecular Weight:
405.87
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

drug control

Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol (± 5%)
100 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CN1CC[C@]23[C@H]4Oc5c(OC(C)=O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4OC(C)=O)c35

InChI

1S/C21H23NO5.ClH/c1-11(23)25-16-6-4-13-10-15-14-5-7-17(26-12(2)24)20-21(14,8-9-22(15)3)18(13)19(16)27-20;/h4-7,14-15,17,20H,8-10H2,1-3H3;1H/t14-,15+,17-,20-,21-;/m0./s1

InChI key

FZJYQGFGNHGSFX-PVQKIFDLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Heroin hydrochloride solution may be used as a reference standard for the analysis of heroin in plasma samples using high-performance liquid chromatography (HPLC) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The quantitative analysis of heroin, methadone and their metabolites and the simultaneous detection of cocaine, acetylcodeine and their metabolites in human plasma by high-performance liquid chromatography coupled with tandem mass spectrometry.
Rook.JE, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 824(1-2), 213-221 (2005)
Ruixue Yin et al.
International journal of medical sciences, 10(3), 299-306 (2013-02-21)
To investigate whether microvascular damage is involved in the pathogenesis of heroin induced spongiform leukoencephalopathy (HSLE). The brain tissues were collected from 4 HSLE patients and 5 controls and then fixed in 4% paraformaldehyde. The frontal lobe, corpus callosum and
Marc Walter et al.
Journal of clinical psychopharmacology, 33(2), 193-198 (2013-02-21)
Heroin dependence is associated with a stressful environment and with dysfunction of the hypothalamic-pituitary-adrenal (HPA) axis. The present study examined the acute effects of intravenous heroin versus placebo on the HPA axis response in heroin-dependent patients. Twenty-eight heroin-dependent patients in
I Demaret et al.
Revue medicale de Liege, 68(5-6), 287-293 (2013-07-31)
Heroin (or diacetylmorphine), a depressant nervous central system, is a semi-synthetic opiate. Its main adverse effect, respiratory depression, can lead to death, especially after an intravenous injection. By loss of tolerance, an overdose can be lethal following heroin use after
D Adam Algren et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 9(1), 106-115 (2013-01-30)
During the summer of 2005, multiple cities in the United States began to report outbreaks of fentanyl-associated fatalities among illicit drug users. The objectives of this study were to (1) determine if an outbreak of fentanyl-associated fatalities occurred in mid-2005

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service