Skip to Content
Merck
All Photos(1)

Key Documents

CRM962

Supelco

PCB Congeners - Loamy Sand

pkg of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM962-050

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    2,2',3,4,4',5,5'-Heptachlorobiphenyl
    2,2′,3,4,4′,5′-Hexachlorobiphenyl
    2,2′,4,4′,5,5′-Hexachlorobiphenyl
    2,2′,4,5,5′-Pentachlorobiphenyl
    2,2′,5,5′-Tetrachlorobiphenyl
    2,3,3′,4,4′,5-Hexachlorobiphenyl
    2,3,3′,4,4′-Pentachlorobiphenyl
    2,3′,4,4′,5-Pentachlorobiphenyl
    2,4,4′-Trichlorobiphenyl
    3,3′,4,4′-Tetrachlorobiphenyl
    See All (10)

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service