Skip to Content
Merck
All Photos(1)

Key Documents

CLNSOIL3

Supelco

Clean Sandy Loam

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€344.00

€344.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€344.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€344.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC CLNSOIL-3-100

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-30°C

Application

Product is intended for use as a clean control and as a base for fortification when no suitable CRM is available for use in method validation, method development or matrix recovery applications. COA covers most paramaters of interest.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The impact of contact time on pyrene sorptive behavior by a sandy-loam soil.
Hwang S and Cutright J T.
Environmental Pollution (Barking, Essex : 1987), 117(3), 371-378 (2002)

Questions

  1. Bonjour, Je souhaiterai savoir si la référence CLNSOIL3 correspond à de la terre stérile ? et si oui, par quelle méthode : autoclavage ? Merci

    1 answer
    1. This product is not sterile. It is intended to be used as a soil sample matrix, with certified levels of impurities documented on the certificate of analysis (COA).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service