Skip to Content
Merck
All Photos(1)

Documents

91588

Supelco

N-(n-Butyl)thiophosphoric triamide

analytical standard

Synonym(s):

N-Butylphosphorothioic triamide, N-Butylthiophosphoric triamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H14N3PS
CAS Number:
Molecular Weight:
167.21
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water (calculated from elemental analysis)

mp

56-58 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

S=P(NCCCC)(N)N

InChI

1S/C4H14N3PS/c1-2-3-4-7-8(5,6)9/h2-4H2,1H3,(H5,5,6,7,9)

InChI key

HEPPIYNOUFWEPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

N-(n-Butyl)thiophosphoric triamide (nBPTT), an active ingredient in nitrogen stabilizers, is widely used as a urease inhibitor. Its mode of action involves the inhibition of urease enzyme thereby improving the efficiency of urea-containing fertilizers. nBPTT is also effective in increasing plant yields and reducing ammonia losses when employed with surface-applied urea and urea ammonium nitrate solutions.

Application

N-(n-Butyl)thiophosphoric triamide may be used as an analytical reference standard for the quantification of the analyte in soil samples, bovine milk and tissue extracts using chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC Method for the Analysis of the Urease Inhibitor N-(n-butyl) thiophosphoric Triamide and Its Metabolites
Douglass AE and Hendrickson LL
Journal of Agricultural and Food Chemistry, 39(12), 2318-2321 (1991)
Nondetectable or minimal detectable residue levels of N-(n-butyl) thiophosphoric triamide in bovine tissues and milk from a 28-d NBPT dosing study
Ligt dVJ, et al.
Translational behavioral medicine, 3(4), 1606-1616 (2019)
Simon Svane et al.
Scientific reports, 10(1), 8503-8503 (2020-05-24)
The nickel-dependent urease enzyme is responsible for the hydrolysis of urea to ammonia and carbon dioxide. A number of bacteria produce urease (ureolytic bacteria) and are associated with various infectious diseases and ammonia emissions from agriculture. We report the first
Lindsay M LaFave et al.
Cancer cell, 38(2), 212-228 (2020-07-25)
Regulatory networks that maintain functional, differentiated cell states are often dysregulated in tumor development. Here, we use single-cell epigenomics to profile chromatin state transitions in a mouse model of lung adenocarcinoma (LUAD). We identify an epigenomic continuum representing loss of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service