Skip to Content
Merck
All Photos(1)

Key Documents

91404

Supelco

3-Hydroxytyrosol

analytical standard

Synonym(s):

2-(3,4-Dihydroxyphenyl)ethanol, 3,4-Dihydroxyphenethyl alcohol, DOPET, Homoprotocatechuyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€97.00

€97.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€97.00

About This Item

Empirical Formula (Hill Notation):
C8H10O3
CAS Number:
Molecular Weight:
154.16
Beilstein:
2208118
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€97.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

OCCc1ccc(O)c(O)c1

InChI

1S/C8H10O3/c9-4-3-6-1-2-7(10)8(11)5-6/h1-2,5,9-11H,3-4H2

InChI key

JUUBCHWRXWPFFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Metabolite of oleuropein. Antioxidant. Inhibits the rate of cancer cell proliferation and induces cancer cell apoptosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service