Skip to Content
Merck
All Photos(2)

Documents

50793

Supelco

Punicalagin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H28O30
CAS Number:
Molecular Weight:
1084.72
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥85.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

OC1O[C@@H]2COC(=O)c3cc(O)c(O)c(O)c3-c4c(O)c(O)c5OC(=O)c6c(c(O)c(O)c7OC(=O)c4c5-c67)-c8c(O)c(O)c(O)cc8C(=O)O[C@H]2[C@@H]9OC(=O)c%10cc(O)c(O)c(O)c%10-c%11c(O)c(O)c(O)cc%11C(=O)O[C@@H]19

InChI

1S/C48H28O30/c49-10-1-6-17(31(59)27(10)55)19-23-21-22-24(47(70)76-38(21)35(63)33(19)61)20(34(62)36(64)39(22)75-46(23)69)18-9(4-13(52)28(56)32(18)60)43(66)74-37-14(5-72-42(6)65)73-48(71)41-40(37)77-44(67)7-2-11(50)25(53)29(57)15(7)16-8(45(68)78-41)3-12(51)26(54)30(16)58/h1-4,14,37,40-41,48-64,71H,5H2/t14-,37-,40+,41-,48?/m1/s1

InChI key

ZJVUMAFASBFUBG-UYMKNUMKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Punicalagin is a water soluble ellagitannin found in pomegranate juice with high antioxidant activity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Potent immune suppressant isolated from pomegranate.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Evaluation of the bioavailability and metabolism in the rat of punicalagin, an antioxidant polyphenol from pomegranate juice.
Cerda B, et al.
European Journal of Nutrition, 42(1), 18-28 (2003)
Repeated oral administration of high doses of the pomegranate ellagitannin punicalagin to rats for 37 days is not toxic.
Cerda B, et al.
Journal of Agricultural and Food Chemistry, 51(11), 3493-3501 (2003)
Dobroslawa Bialonska et al.
Journal of agricultural and food chemistry, 57(18), 8344-8349 (2009-08-27)
The consumption of pomegranate products leads to a significant accumulation of ellagitannins in the large intestines, where they interact with complex gut microflora. This study investigated the effect of pomegranate tannin constituents on the growth of various species of human
Anna Carreras et al.
Journal of agricultural and food chemistry, 60(7), 1659-1665 (2012-01-28)
Plant polyphenols may be free radical scavengers or generators, depending on their nature and concentration. This dual effect, mediated by electron transfer reactions, may contribute to their influence on cell viability. This study used two stable radicals (tris(2,3,5,6-tetrachloro-4-nitrophenyl)methyl (TNPTM) and
Sylvie Hollebeeck et al.
Food & function, 3(8), 875-885 (2012-06-27)
This study aimed at evaluating the anti-inflammatory properties of a pomegranate fruit husk (PomH) polyphenolic extract, rich in punicalagin, using Caco-2 cells, an in vitro model of human intestinal epithelium. Differentiated cells in bicameral inserts were pretreated or not with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service