Skip to Content
Merck
All Photos(1)

Key Documents

89257

Supelco

Fenthion oxon

PESTANAL®, analytical standard

Synonym(s):

O,O′-Dimethyl O′′-[3-methyl-4-(methylthio)phenyl] phosphate, Dimethyl 4-(methylthio)-m-tolyl phosphate, Fenoxon, Fenthoxon

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€99.10

€99.10


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€99.10

About This Item

Empirical Formula (Hill Notation):
C10H15O4PS
CAS Number:
Molecular Weight:
262.26
Beilstein:
2275764
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€99.10


Estimated to ship on21 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

O=P(OC1=CC(C)=C(SC)C=C1)(OC)OC

InChI

1S/C10H15O4PS/c1-8-7-9(5-6-10(8)16-4)14-15(11,12-2)13-3/h5-7H,1-4H3

InChI key

ZNRZGJAHNMGWQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fenthion oxon may be used as a reference standard for the determination of fenthion oxon in animal tissue samples by gel permeation chromatography (GPC) cleanup followed by gas chromatography-mass spectrometry (GC–MS) and liquid chromatography coupled with tandem mass spectrometry (LC–MS–MS).[1]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation study on 660 pesticide residues in animal tissues by gel permeation chromatography cleanup/gas chromatography-mass spectrometry and liquid chromatography-tandem mass spectrometry.
Pang GF, et al.
Journal of Chromatography A, 1125(1), 1-30 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service