Skip to Content
Merck
All Photos(1)

Key Documents

89176

Millipore

Muller-Kauffmann Tetrathionate Novobiocin Broth

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€99.60

€99.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€99.60

About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

€99.60


Check Cart for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

brilliant green, 0.0095 g/L
calcium carbonate, 38.7 g/L
Casein enzymic hydrolysate, 8.6 g/L
ox bile, 4.75 g/L
peptic digest of animal tissue, 4.3 g/L
sodium chloride, 2.6 g/L
sodium thiosulphate, pentahydrate, 47.8 g/mL

manufacturer/tradename

NutriSelect® Plus

final pH

8.2±0.2 (25 °C)

application(s)

environmental
food and beverages
sample preparation

microbiology

suitability

Salmonella spp.

Application

Mueller Kauffman Tetrathionate Novobiocin Broth is used for improved enrichment and isolation of Salmonellae.

Preparation Note

Suspend 89.42 grams of dehydrated mediumI in 1000 ml distilled water. Heat the medium just to boiling. DO NOT AUTOCLAVE. Cool to 45-50°C and just before use aseptically add 20 ml of iodine solution (20 gram iodine and 25 gram potassium iodide in 100 ml sterile distilled water) along with rehydrated contents of 1 vial of MKTT Novobiocin Supplement (Cat. No. 79894). Mix well to disperse calcium carbonate uniformly before dispensing in sterile tubes.
Note: Due to presence of calcium carbonate, the prepared media forms opalescent solution with white precipitate.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service