Skip to Content
Merck
All Photos(1)

Documents

79894

Millipore

MKTTn Novobiocin Supplement

suitable for microbiology

Synonym(s):

Muller-Kauffmann Tetrathionate Novobiocin Supplement, Novobiocin Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Salmonella spp.

Related Categories

Application

Supplement recommended for selective enrichment and isolation of Salmonellae.

Analysis Note

(per vial sufficient for 1000 ml medium)
Novobiocin 40.00mg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service