Skip to Content
Merck
All Photos(1)

Key Documents

88975

Sigma-Aldrich

Bicarbonate buffer solution

BioUltra, >0.1 M Na2CO3, >0.2 M NaHCO3

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€32.30

€32.30


Estimated to ship on19 March 2025


Request a Bulk Order

Select a Size

Change View
10 ML
€32.30

About This Item

MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

€32.30


Estimated to ship on19 March 2025


Request a Bulk Order

product line

BioUltra

Quality Level

form

liquid

concentration

>0.1 M Na2CO3
>0.2 M NaHCO3

impurities

insoluble matters, passes filter test
metal traces

pH

9.5±0.2

density

1.02 g/mL at 20 °C

λ

neat

UV absorption

λ: 260 nm Amax: <0.02
λ: 280 nm Amax: <0.02

Application

Bicarbonate buffer solution is a physiological buffer used in the preparation of a wide range of cell and molecular biology reagent solutions.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola J Robertson et al.
Pediatric research, 58(2), 288-296 (2005-07-12)
A period of secondary energy failure consisting of a decline in phosphocreatine/inorganic phosphate (PCr/Pi), a rise in brain lactate, and alkaline intracellular pH (pH(i)) has been described in infants with neonatal encephalopathy. Strategies that ameliorate this energy failure may be
T G Ramsay et al.
Journal of animal science, 83(11), 2501-2508 (2005-10-19)
The present study was performed to examine the response of leptin and leptin receptor (Rb) genes to porcine somatotropin (pST) stimuli in finishing pigs. Twelve crossbred barrows (Yorkshire x Landrace) were used in this study. Animals were individually fed a
Visualizing dengue virus through Alexa Fluor labeling.
Zhang S, Tan HC, Ooi EE.
Journal of Visualized Experiments, 9, e3168-e3168 (2011)
Christopher O Phillips et al.
Langmuir : the ACS journal of surfaces and colloids, 28(25), 9878-9884 (2012-05-24)
Antibodies were patterned onto flexible plastic films using the flexographic printing process. An ink formulation was developed using high molecular weight polyvinyl alcohol in carbonate-bicarbonate buffer. In order to aid both antibody adhesion and the quality of definition in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service