Skip to Content
Merck

Skip To

1.01962

Sodium hydrogen carbonate/sodium carbonate

certified reference material

Synonym(s):

Sodium Carbonate/Na2CO3, Sodium bicarbonate/NaHCO3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 x 25 g
Check Cart for Availability
€ 367,00
€ 275,25

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

€ 275,25

List Price€ 367,00Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

complies with DIN 19266

product line

Certipur®

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

neat

storage temp.

15-25°C

General description

Certipur® certified secondary standard reference buffers are used to precisely calibrate and monitor pH measurement instruments and buffer solutions. They are quality controlled and qualified in our ISO/IEC 17025 and 17034 accredited calibration laboratories against certified primary standard reference materials (SRM) in compliance with DIN 19266, IUPAC, NIST, Ph. Eur. and USP. Certipur® reference materials offer maximum accuracy, reliability and convenience and help avoid costly repeat analyses.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Sodium hydrogen carbonate/sodium carbonate [pH(S) = 10.012 (25°C)] is a certified secondary standard reference buffer used as a calibration buffer standard for pH instruments or pH electrodes.

Features and Benefits

  • The product is delivered with a comprehensive, batch-specific Certificate of Analysis showing relevant data for the quality management system.
  • The pH value of this Certipur® buffer solution is measured using differential potentiometry with two platinum hydrogen electrodes against NIST & PTB primary SRM [pH(S) = 10.012 (25 °C)].
  • It is a Certified Reference Material (CRM) offered in neat format.
  • Intended for extremely precise calibration, qualification and monitoring of pH instruments and for testing other buffer solutions.

Analysis Note

pH-value (25 °C): 10.010 - 10.016

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.019641.01963PHR1948
format

neat

format

neat

format

neat

format

-

product line

Certipur®

product line

Certipur®

product line

Certipur®

product line

-

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material, pharmaceutical secondary standard

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

pharmaceutical

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

2-8°C

agency

complies with DIN 19266

agency

complies with DIN 19266

agency

complies with DIN 19266

agency

traceable to USP 1613757


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters