Skip to Content
Merck
All Photos(1)

Documents

87894

Supelco

α-Amylcinnamyl alcohol

analytical standard

Synonym(s):

2-Benzylidene-1-heptanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C[(CH2)4CH3]CH2OH
CAS Number:
Molecular Weight:
204.31
Beilstein:
3127316
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.519 (lit.)
n20/D 1.537

bp

141-143 °C/5 mmHg (lit.)

density

0.952 g/mL at 25 °C (lit.)
0.966 g/mL at 25 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCC\C(CO)=C/c1ccccc1

InChI

1S/C14H20O/c1-2-3-5-10-14(12-15)11-13-8-6-4-7-9-13/h4,6-9,11,15H,2-3,5,10,12H2,1H3/b14-11+

InChI key

LIPHCKNQPJXUQF-SDNWHVSQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S32-S39 (2007-11-27)
A toxicologic and dermatologic review of alpha-amylcinnamyl alcohol when used as a fragrance ingredient is presented.
J Ashby et al.
Carcinogenesis, 3(11), 1273-1276 (1982-01-01)
The rate of hydrolysis to formaldehyde of methylenedimorpholine (MDM), hexamethylenetetramine (HMT) and dinitrosopentamethylenetetramine (DNPT) have been compared with the enzyme-mediated formation of formaldehyde from hexamethylphosphoramide (HMPA). The bio-distribution of [14C]HMPA following nasal instillation in rats has also been studied and
J D Guin et al.
Journal of the American Academy of Dermatology, 8(1), 76-80 (1983-01-01)
Sensitivity to alpha-amylcinnamic aldehyde (alpha-AcAld) is apparently uncommon, but, like allergy to alpha-amylcinnamic alcohol (alpha-AcAlc), it often accompanies allergy to the perfume in Mycolog cream. Although alpha-AcAlc is a known ingredient, alpha-AcAld is not. However, gas-liquid chromatographic analysis shows alpha-AcAld

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service