Skip to Content
Merck
All Photos(1)

Key Documents

693017

Millipore

Zwittergent® 3-14 Detergent

Synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a wide pH range.

Synonym(s):

n-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, hplc solvents

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€201.00
25 G
€684.00
100 G
€2,170.00
500 G
€3,530.00

€201.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€201.00
25 G
€684.00
100 G
€2,170.00
500 G
€3,530.00

About This Item

Empirical Formula (Hill Notation):
C19H41NO3S
CAS Number:
Molecular Weight:
363.60
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25

€201.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥99% (TLC)

form

powder
solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

aggregation number

83

color

white

CMC

100 - 400 μM

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])CCC[N+](CCCCCCCCCCCCCC)(C)C

InChI

1S/C19H41NO3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-17-20(2,3)18-16-19-24(21,22)23/h4-19H2,1-3H3

InChI key

BHATUINFZWUDIX-UHFFFAOYSA-N

General description

Synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a wide pH range. This property is attributed to the presence of both a strongly basic quaternary ammonium ion and an acidic sulfonate ion of equal strength. This ionic balance presumably prevents irreversible binding of the detergent to either anionic or cationic compounds.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, refrigerate (4°C). Aqueous stock solutions are stable for several months at 4°C.

Other Notes

Aveldano, M.I., et al. 1991. J. Neurochem.57, 250.
Wach, A., et al. 1990. Eur. J. Biochem.189, 675.
Lai, F.A., et al. 1989. J. Biol. Chem. 264, 16776.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Zwittergent is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service