Skip to Content
Merck
All Photos(1)

Key Documents

85973

Supelco

Methoxypolyethylene glycol 5,000 acetic acid N-succinimidyl ester

≥80%

Synonym(s):

Polyethylene glycol, mono-Methyl polyethylene glycol 5′000 acetic acid N-succinimidyl ester, Succinimidyl ester of carboxymethyl-PEG

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€524.00

€524.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€524.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

€524.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥80% (NMR)
≥80%

form

powder

mol wt

PEG average Mn 5,000

Ω-end

NHS ester

α-end

methoxy

shipped in

wet ice

storage temp.

−20°C

SMILES string

N1(C(=O)CCC1=O)OC(=O)COCCO.O(CCO)C

InChI

1S/C8H11NO6.C3H8O2/c10-3-4-14-5-8(13)15-9-6(11)1-2-7(9)12;1-5-3-2-4/h10H,1-5H2;4H,2-3H2,1H3

InChI key

NYXRHTOXAYVBMV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service