Skip to Content
Merck
All Photos(1)

Key Documents

81330

Sigma-Aldrich

Poly(methylhydrosiloxane)

viscosity 15-40 mPa.s (20 °C)

Synonym(s):

Methylhydrogensiloxane polymer, PMHS

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€72.60
500 ML
€201.00

€72.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€72.60
500 ML
€201.00

About This Item

Linear Formula:
(CH3)3SiO[(CH3)HSiO]nSi(CH3)3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€72.60


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

refractive index

n20/D 1.398

viscosity

15-40 mPa.s(20 °C)

density

1.006 g/mL at 25 °C (lit.)

SMILES string

[Si](C)(C)(C)C.[Si](O)(C)(C)C.[SiH](O)(C)C

InChI key

KZFYHEHDBPYRMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reducing agent[1][2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

399.2 °F - closed cup

Flash Point(C)

204 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Chuit et al.
Synthesis, 981-981 (1982)
I. Pri-Bar et al.
The Journal of Organic Chemistry, 51, 734-734 (1986)

Questions

  1. Is additional curing (cross-linking) chemical needed for product 81330, Poly(methylhydrosiloxane)?

    1 answer
    1. product 81330 does not cross-link on its own at temperatures below 200°C. It requires a curing agent, with the most common options being basic materials such as amines, metal oxides, and metal hydroxides. However, caution is advised during the cross-linking process, as too rapid a cross-link can lead to rapid hydrogen gas evolution, which is highly flammable. This risk can be mitigated by using a lower dosing of the base or a weaker basic material.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service