Skip to Content
Merck
All Photos(2)

Key Documents

8.18063

Sigma-Aldrich

Poly(methyl hydrogen siloxane)

for synthesis

Synonym(s):

Poly(methyl hydrogen siloxane), PMHS

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€75.20
1 L
€151.00

€75.20


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€75.20
1 L
€151.00

About This Item

Linear Formula:
(CH4OSi)nC6H18OSi2
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

€75.20


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

38 hPa ( 20 °C)

Quality Level

form

liquid

reaction suitability

reagent type: reductant

pH

7 ( in H2O)

kinematic viscosity

17-25 cSt(25 °C)

transition temp

flash point 150 °C

density

1.00 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](C)(C)(C)C.[Si](O)(C)(C)C.[SiH](O)(C)C

InChI

1S/C4H12Si.C3H10OSi.C2H8OSi/c2*1-5(2,3)4;1-4(2)3/h1-4H3;4H,1-3H3;3-4H,1-2H3

InChI key

KZFYHEHDBPYRMU-UHFFFAOYSA-N

Application

  • Development of superhydrophobic coatings: Research conducted in 2019 utilized Poly(methyl hydrogen siloxane) for creating superhydrophobic coatings by the in situ growth of silicone nanofilaments on alkali-activated geopolymers. This application is critical for developing moisture-resistant coatings in construction and exterior surface treatments to increase longevity and resistance to environmental factors (Liu et al., 2019).

Analysis Note

Density (d 20 °C/ 4 °C): 0.995 - 1.005
Hydrogen releasable in alkaline medium: ≥ 340 ml/g
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

399.2 °F - closed cup

Flash Point(C)

204 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service